Learn More
Sigma Aldrich Fine Chemicals Biosciences GABase from Pseudomonas fl
Catalog No. 502904637
Encompass
GABase is a commercially available mixture composed of³-aminobutyric acid aminotransferase (GABGT) and succinic semialdehyde dehydrogenase (SSDH) obtained from Pseudomonas fluorescens.
Catalog No. 50-290-4637 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G750910UN
GABase is a commercially available mixture composed of³-aminobutyric acid aminotransferase (GABGT) and succinic semialdehyde dehydrogenase (SSDH) obtained from Pseudomonas fluorescens.