Learn More
Medchemexpress LLC HY-103533 5mg Medchemexpress, Gabazine CAS:104104-50-9 Purity:>98%
SDP
Catalog No. 501968126
Encompass_Preferred
Medchemexpress, HY-103533 5mg Gabazine CAS:104104-50-9 Gabazine is a selective and competitive antagonist of GABAA receptor, with an IC50 of ~0.2 μM for GABA receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8126 Supplier Medchemexpress LLC Supplier No. HY1035335MG
Medchemexpress, HY-103533 5mg Gabazine CAS:104104-50-9 Gabazine is a selective and competitive antagonist of GABAA receptor, with an IC50 of ~0.2 μM for GABA receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.