Learn More
Aviva Systems Biology Corp GABBR2 Antibody - C-terminal region (ARP61115_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61115_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GABBR2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human GABBR2. Alternate names: FLJ36928, GABABR2, GPR51, GPRC3B, HG20, HRIHFB2099ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-287-257