Learn More
Selleck Chemical LLC Gabexate Mesylate 5mg 56974-61-9
SDP
Supplier:  Selleck Chemical LLC S21015MG
Encompass_Preferred
Gabexate Mesylate is a serine protease inhibitor with IC50 of 0.19 ?M which is used therapeutically in the treatment of pancreatitis and disseminated intravascular coagulation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1764304