Learn More
Medchemexpress LLC Gabexate (mesylate) | 56974-61-9 | 98.64% | 100 MG
SDP
Catalog No. 5000407159
Encompass_Preferred
Gabexate mesylate (FOY) is a competitive and non-antigenic synthetic inhibitor of trypsin-like serine proteinases. It inhibits human thrombin, urokinase, plasmin, and Factor Xa, and binds to human and bovine tryptase. It is used for pancreatitis and disseminated intravascular coagulation, and decreases proliferation rate in HepG2 cells in vitro.
  • Anticoagulant effect by inhibiting the clotting activity of thrombin.
  • Anti-inflammatory effect through the inhibition of NF-κB, proinflammatory cytokines, and nitric oxide.
  • Therapeutic applications for pancreatitis and disseminated intravascular coagulation.
  • Decreases the proliferation rate of HepG2 cells in a dose-dependent manner.
  • Attenuates the development of mechanical allodynia induced by spinal nerve ligation (SNL) and inhibits the expression of pro-inflammatory cytokines after SNL in a rat model.
Catalog No. 50-004-07159 Supplier Medchemexpress LLC Supplier No. HYB0385100MG
Add to Cart
Add to Cart
Gabexate mesylate (FOY) is a competitive and non-antigenic synthetic inhibitor of trypsin-like serine proteinases. It inhibits human thrombin, urokinase, plasmin, and Factor Xa, and binds to human and bovine tryptase. It is used for pancreatitis and disseminated intravascular coagulation, and decreases proliferation rate in HepG2 cells in vitro.
  • Anticoagulant effect by inhibiting the clotting activity of thrombin.
  • Anti-inflammatory effect through the inhibition of NF-κB, proinflammatory cytokines, and nitric oxide.
  • Therapeutic applications for pancreatitis and disseminated intravascular coagulation.
  • Decreases the proliferation rate of HepG2 cells in a dose-dependent manner.
  • Attenuates the development of mechanical allodynia induced by spinal nerve ligation (SNL) and inhibits the expression of pro-inflammatory cytokines after SNL in a rat model.