Learn More
Apexbio Technology LLC Gabexate mesylate, 5mg. Cas: 56974-61-9 MFCD: MFCD00210299. Trypsin-like serine proteinases inhibitor
SDP
Supplier:  Apexbio Technology LLC A40125
Encompass_Preferred
Gabexate mesylate is a non-antigenic synthetic inhibitor of trypsin-like serine proteinases with Ki values of 3.4nM and 180nM, respectively for human tryptase and bovine tryptase [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4853