Learn More
Medchemexpress LLC Gabexate mesylate (FOY) | 56974-61-9 | 98.6% | 50 MG
SDP
Catalog No. 5000407160
Encompass_Preferred
Gabexate mesylate (FOY) is a competitive and non-antigenic synthetic inhibitor of trypsin-like serine proteinases. It exerts an anticoagulant effect on the clotting activity of thrombin and has anti-inflammatory effects. It is used for pancreatitis and disseminated intravascular coagulation.
  • Inhibits human thrombin, urokinase, plasmin, and Factor Xa
  • Binds to human and bovine tryptase
  • Attenuates mechanical allodynia in rats
  • Decreases HepG2 cell proliferation rate
Catalog No. 50-004-07160 Supplier Medchemexpress LLC Supplier No. HYB038550MG
Gabexate mesylate (FOY) is a competitive and non-antigenic synthetic inhibitor of trypsin-like serine proteinases. It exerts an anticoagulant effect on the clotting activity of thrombin and has anti-inflammatory effects. It is used for pancreatitis and disseminated intravascular coagulation.
  • Inhibits human thrombin, urokinase, plasmin, and Factor Xa
  • Binds to human and bovine tryptase
  • Attenuates mechanical allodynia in rats
  • Decreases HepG2 cell proliferation rate