Learn More
Abcam Gabe x ate mesylate, General serine protease inhibitor, 50MG
Catalog No. 502706841
Encompass_Preferred
MW 417.5 Da, Purity >99%. General serine protease inhibitor (Ki values are 3.4 nM, 5, 1.8 μM and 2.6 mM at tryptase, thrombin, tyrpsin and kallikrein serine proteases respectively). Additionally inhibits nitric oxide synthase. Displays antithrombotic activity and antitumor effect. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6841 Supplier Abcam Supplier No. AB14116750MG
MW 417.5 Da, Purity >99%. General serine protease inhibitor (Ki values are 3.4 nM, 5, 1.8 μM and 2.6 mM at tryptase, thrombin, tyrpsin and kallikrein serine proteases respectively). Additionally inhibits nitric oxide synthase. Displays antithrombotic activity and antitumor effect. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement