Learn More
Medchemexpress LLC HY-10233 10mg Medchemexpress, Gaboxadol (hydrochloride) CAS:85118-33-8 Purity:>98%
SDP
Catalog No. 502080653
Encompass_Preferred
Medchemexpress, HY-10233 10mg Gaboxadol (hydrochloride) CAS:85118-33-8 Gaboxadol hydrochloride (Lu 02-030 hydrochloride; THIP hydrochloride) is a potent agonist of the GABAA receptor and an antagonist of GABAC receptors (IC50=25 μM). Gaboxadol hydrochloride displays a partial agonist efficacy on subunit α1β2γ2 with an ED50 value of 143 µM, a full agonist efficacy at α5 subunit (ED50=28-129 µM) and a superagonist efficacy at α4β3δ (ED50=6 µM). Gaboxadol hydrochloride is a non-opioid analgesic and has hypnotic activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0653 Supplier Medchemexpress LLC Supplier No. HY1023310MG
Medchemexpress, HY-10233 10mg Gaboxadol (hydrochloride) CAS:85118-33-8 Gaboxadol hydrochloride (Lu 02-030 hydrochloride; THIP hydrochloride) is a potent agonist of the GABAA receptor and an antagonist of GABAC receptors (IC50=25 μM). Gaboxadol hydrochloride displays a partial agonist efficacy on subunit α1β2γ2 with an ED50 value of 143 µM, a full agonist efficacy at α5 subunit (ED50=28-129 µM) and a superagonist efficacy at α4β3δ (ED50=6 µM). Gaboxadol hydrochloride is a non-opioid analgesic and has hypnotic activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.