Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Gaboxadol hydrochloride

Catalog No. T10125MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10125MG 25 mg
50-290-7697 500 mg
T101100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T10125MG Supplier Sigma Aldrich Supplier No. T10125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Gaboxadol, also referred to as 4,5,6,7-tetrahydroisoxazolo(5,4-c)pyridin-3-ol (thip), is a gaba site agonist
  • It has great affinity to the α subunit of extrasynaptic high-affinity gabaa receptors
  • Gaboxadol was first developed for the treatment of schizophrenia, epilepsy, primary insomnia and huntington's disease
  • It can easily cross the blood-brain barrier and is rapidly absorbed by the body.gaboxadol hydrochloride has been used in cell migration and invasion assay to assess liver cancer cell migration.gabaa receptor agonist

Specifications

CAS 85118-33-8
Molecular Formula C6H8N2O2 -+ HCl
Linear Formula C6H8N2O2 -+ HCl
MDL Number MFCD00055180
Quantity 25 mg
Synonym 4,5,6,7-Tetrahydroisoxazolo[5,4-c]pyridin-3-ol hydrochloride; THIP hydrochloride
Molecular Weight (g/mol) 176.6
Percent Purity ≥98% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.