Learn More
Medchemexpress LLC Gaboxadol hydrochloride | 85118-33-8 | MFCD00055180 | 99.9% | 176.60 g/mol | C6H9ClN2O2 | 1 ML
SDP
Catalog No. 5000246569
Encompass_Preferred
Gaboxadol hydrochloride is the hydrochloride salt of gaboxadol, a GABAA receptor agonist used as a research reagent. This item is supplied as a ready-to-use 10 mM stock solution (1 mL) in DMSO for in vitro research applications.
  • Ready-to-use 10 mM stock solution in DMSO.
  • High purity, 99.85%.
  • Hydrochloride salt; molecular formula C6H9ClN2O2 and molecular weight 176.60 g/mol.
  • Store stock solution at -80°C for long-term storage or -20°C for short-term use.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-46569 Supplier Medchemexpress LLC Supplier No. HY1023310MM 1ML
Gaboxadol hydrochloride is the hydrochloride salt of gaboxadol, a GABAA receptor agonist used as a research reagent. This item is supplied as a ready-to-use 10 mM stock solution (1 mL) in DMSO for in vitro research applications.
  • Ready-to-use 10 mM stock solution in DMSO.
  • High purity, 99.85%.
  • Hydrochloride salt; molecular formula C6H9ClN2O2 and molecular weight 176.60 g/mol.
  • Store stock solution at -80°C for long-term storage or -20°C for short-term use.
  • Intended for research use only; not for human or veterinary use.