Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gaboxadol Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. G040525MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G040525MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G040525MG Supplier TCI America Supplier No. G040525MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 85118-33-8
Molecular Formula C6H9ClN2O2
Molecular Weight (g/mol) 176.60
MDL Number MFCD00055180
InChI Key ZDZDSZQYRBZPNN-UHFFFAOYSA-N
Synonym thip hydrochloride, gaboxadol hydrochloride, unii-478rvh3tvd, 478rvh3tvd, 4,5,6,7-tetrahydroisoxazolo 5,4-c pyridin-3-ol hydrochloride, thip nieani, thip hcl, thip, d00xny, spectrum1503648
PubChem CID 5702253
IUPAC Name 2H,3H,4H,5H,6H,7H-[1,2]oxazolo[5,4-c]pyridin-3-one hydrochloride
SMILES Cl.O=C1NOC2=C1CCNC2

Specifications

Melting Point 236°C
Color White
Quantity 25 mg
Formula Weight 176.60
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Gaboxadol Hydrochloride
RTECSNumber NY3398000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.