Learn More
Abcam Gabo x adol hydrochloride (THIP hydrochloride), GABAA receptor agonist and GABAC receptor antagonist, 25MG
Catalog No. 502707596
Encompass_Preferred
MW 176.6 Da, Purity >98%. GABAA receptor agonist and GABAC receptor antagonist. Antinociceptive, anticonvulsant and hypnotic agent. Blood-brain barrier permeable. Active *in vivo* and *in vitro*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7596 Supplier Abcam Supplier No. AB12042625MG
MW 176.6 Da, Purity >98%. GABAA receptor agonist and GABAC receptor antagonist. Antinociceptive, anticonvulsant and hypnotic agent. Blood-brain barrier permeable. Active *in vivo* and *in vitro*.
The product is subject to the following: Abcam Restricted Use Statement