Learn More
Aviva Systems Biology Corp GABPB2 Antibody - C-terminal region (P100925_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100925_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GABPB2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human GABPB2. Alternate names: E4TF1, GABPB, BABPB2, E4TF1B, GABPB2, NRF2B1, NRF2B2, E4TF1-47, E4TF1-53ApplicationWB. Species reactivity:
Catalog No. 50-014-62786