Learn More
Aviva Systems Biology Corp GABPB2 Antibody - middle region (ARP31189_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31189_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GABPB2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GABPB2. Alternate names: BABPB2, E4TF1, E4TF1-47, E4TF1-53, E4TF1B, GABPB, GABPB1, NRF2B1, NRF2B2, GABPB2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54184