Learn More
ABclonal Technology GABRA5 Rabbit pAb
SDP
Supplier:  ABclonal Technology A139841000UL
Encompass_Preferred
GABA is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor. At least 16 distinct subunits of GABA-A receptors have been identified. Transcript variants utilizing three different alternative non-coding first exons have been described.
Catalog No. 50-001-09318