Learn More
Aviva Systems Biology Corp GABRB2 Antibody - N-terminal region (ARP34989_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34989_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GABRB2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GABRB2. Alternate names: ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-52984