Learn More
Aviva Systems Biology Corp GABRE Antibody - N-terminal region (ARP35156_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35156_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GABRE. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GABRE. Alternate names: -ApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Rabbit
Catalog No. 50-014-55654