Learn More
Aviva Systems Biology Corp GABRE Antibody - middle region (ARP35342_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35342_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GABRE. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GABRE. Alternate names: ApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53001