Learn More
ABclonal Technology GABRG1 Rabbit pAb
SDP
Catalog No. 5000106441
Encompass_Preferred
The protein encoded by this gene belongs to the ligand-gated ionic channel family. It is an integral membrane protein and plays an important role in inhibiting neurotransmission by binding to the benzodiazepine receptor and opening an integral chloride channel. This gene is clustered with three other family members on chromosome 4.
Catalog No. 50-001-06441 Supplier ABclonal Technology Supplier No. A14240500UL
The protein encoded by this gene belongs to the ligand-gated ionic channel family. It is an integral membrane protein and plays an important role in inhibiting neurotransmission by binding to the benzodiazepine receptor and opening an integral chloride channel. This gene is clustered with three other family members on chromosome 4.