Learn More
Aviva Systems Biology Corp GABRG2 Antibody - N-terminal region (AVARP13033_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13033_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GABRG2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GABRG2. Alternate names: CAE2, ECA2, GEFSP3ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62617