Learn More
Aviva Systems Biology Corp GABRG3 Antibody - N-terminal region (AVARP13101_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13101_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GABRG3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GABRG3. Alternate names: ApplicationWB. Species reactivity: Human, Mouse
Catalog No. 50-014-62641