Learn More
Aviva Systems Biology Corp GABRQ Antibody - N-terminal region (ARP35283_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35283_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GABRQ. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GABRQ. Alternate names: THETAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-52999