Learn More
Genscript Corporation Antibody Protein Proteins Proteins 10 UG 1/EA
Supplier:  Genscript Corporation Z0311210
Encompass
gAcrp30 is the globular head domain of Adipocyte complement-related protein of 30 kDa (Acrp30) a cytokine expressed in adipocytes. The name of Acrp30 is bases on its closest homolog complement factor c1q and the globular domain of Acrp30 has an unexpected homology with the Tumor Necrosis Factor (TNF) family of cytokines. Acrp30 is recognized by two receptors: adipoR1 expressed in skeletal muscle and adipoR2 expressed in liver. The expression level of Acrp30 in adipocytes is negatively correlated with body weight and is lower in obese mouse than normal mouse. The globular domain of Acrp30 induces free fatty acid oxidation in muscle and weight reduction in mouse suggesting its potential use as a pharmacological agent in obesity.
Catalog No. 50-252-9827