Learn More
Enzo Life Sciences Glutamic acid decarboxylase monoclonal antibody (11) (100µl)
SDP
Supplier:  Enzo Life Sciences BMLGC32080100
Encompass_Preferred
Isotype: IgG1. Immunogen: The hybridoma secreting the antibody to GAD65 was generated by fusion of splenocytes from a non-obese diabetic (NOD) mouse which had received a single subdiabetogenic injection of streptozotocin. Effectively, BML-GC3208 is a mouse autoantibody to GAD. Species reactivity: Human, Mouse, Rat, Monkey. Specificity: Recognizes an N-terminal epitope. The antibody consistently labels neuronal GAD in the CNS and GAD-containing cells in mammalian (not mouse) pancreas. Applications: IHC, WB. Purity: Partially purified. Formulation: Liquid. In PBS containing 10mM sodium azide.
Catalog No. 50-201-0900