Learn More
Enzo Life Sciences Glutamic acid decarboxylase polyclonal antibody (25µl)
SDP
Supplier:  Enzo Life Sciences BMLGC30080025
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 572-585 (D572FLIEEIERLGQDL585) of rat glutamate decarboxylase (GAD65). This sequence is conserved in rat GAD67 and in human GAD-2. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat Cat. Applications: ELISA, IHC, WB. Purity: Protein A-affinity chromatography. Formulation: Liquid. In PBS containing 0.09% sodium azide.
Catalog No. 50-201-0897