Learn More
Aviva Systems Biology Corp Gad1 Antibody - N-terminal region (ARP54354_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54354_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Gad1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human Gad1. Alternate names: EP10, GAD25, GAD44, GAD67, Gad-1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-910