Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GAD65 (524-543) acetate | 152468-44-5 | 98.1% | 2238.67 | 1 MG
SDP

Catalog No. 5000398926
Encompass_Preferred

GAD65 (524-543) acetate | 152468-44-5 | 98.1% | 2238.67 | 1 MG

Catalog No. 50-003-98926 Supplier Medchemexpress LLC Supplier No. HYP5396A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GAD65 (524-543) acetate | 152468-44-5 | 98.1% | 2238.67 | 1 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.