Learn More
Charles River Labs Cell Solutions GAD-65 (555-567) 1mg, Cryopreserved
Supplier:  Charles River Labs Cell Solutions ANTIGEN1135
Encompass
Glutamic acid decarboxylase is an important autoantigen in diabetes. The sequence of this peptide is NFFRMVISNPAAT.
Catalog No. 50-225-6098
Add to Cart
This item is not returnable. View return policy
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov