Learn More
ABclonal Technology GAD65/GAD2 Rabbit mAb
SDP
Catalog No. 5000007302
Encompass_Preferred
This gene encodes one of several forms of glutamic acid decarboxylase identified as a major autoantigen in insulin-dependent diabetes The enzyme encoded is responsible for catalyzing the production of gamma-aminobutyric acid from L-glutamic acid A pathogenic role for this enzyme has been identified in the human pancreas since it has been identified as an autoantibody and an autoreactive T cell target in insulin-dependent diabetes This gene may also play a role in the stiff man syndrome Alternative splicing results in multiple transcript variants that encode the same protein
Catalog No. 50-000-07302 Supplier ABclonal Technology Supplier No. A22728100UL
This gene encodes one of several forms of glutamic acid decarboxylase identified as a major autoantigen in insulin-dependent diabetes The enzyme encoded is responsible for catalyzing the production of gamma-aminobutyric acid from L-glutamic acid A pathogenic role for this enzyme has been identified in the human pancreas since it has been identified as an autoantibody and an autoreactive T cell target in insulin-dependent diabetes This gene may also play a role in the stiff man syndrome Alternative splicing results in multiple transcript variants that encode the same protein