Learn More
Aviva Systems Biology Corp GADD45A Antibody - C-terminal region (ARP60167_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60167_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GADD45A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human GADD45A. Alternate names: DDIT1, GADD45ApplicationWB. Species reactivity: Cow, Horse, Human, Pig, Rabbit
Catalog No. 50-286-968