Learn More
Aviva Systems Biology Corp GADD45B Antibody - middle region (ARP48346_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48346_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GADD45B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GADD45B. Alternate names: DKFZP566B133, GADD45BETA, MYD118ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-53549