Learn More
Selleck Chemical LLC Gadodiamide Hydrate 50mg 122795-43-1
SDP
Catalog No. 501948092
Encompass_Preferred
Gadodiamide Hydrate, a nonionic Gd3+ chelate, is frequently injected i.v. into magnetic resonance imaging (MRI) to enhance contrast. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8092 Supplier Selleck Chemical LLC Supplier No. S175150MG
Gadodiamide Hydrate, a nonionic Gd3+ chelate, is frequently injected i.v. into magnetic resonance imaging (MRI) to enhance contrast. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.