Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gadolinium boride, 99.5% (metals basis)

Catalog No. AA3922503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3922503 1 g
AA3922506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3922503 Supplier Thermo Scientific Chemicals Supplier No. 03922503
Add to Cart

CAS: 12008-06-9 | B6Gd | 222.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12008-06-9
Molecular Formula B6Gd
Molecular Weight (g/mol) 222.11
MDL Number MFCD00049942
InChI Key FYKPSYVUFHLGOY-UHFFFAOYSA-N
Synonym gadolinium boride, gadolinium boride, gdb6, gadolinium 2+ pentacyclo 3.1.0.0^ 1,3 .0^ 2,4 .0^ 4,6 hexaborane-1,4-diuide, gadolinium 2+ ion pentacyclo 3.1.0.0 1 , 3 .0 2 ,?.0?,? hexaborane-1,4-diuide
PubChem CID 25021694
IUPAC Name gadolinium(2+) pentacyclo[3.1.0.0¹,³.0²,⁴.0⁴,⁶]hexaborane-1,4-diuide
SMILES [Gd++].B12B3[B-]11B4B1[B-]234

Specifications

Physical Form Powder
Quantity 1 g
Formula Weight 222.12
Percent Purity 99.5%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Gadolinium boride
EINECSNumber 234-530-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.