Learn More
Strem, An Ascensus Company CAS# 19423-81-5. 100g. Gadolinium(III) chloride hydrate (99.9%-Gd) (REO). MFCD00149731
SDP
Catalog No. 5090116063
Encompass
CAS# 19423-81-5. 100g. Gadolinium(III) chloride hydrate (99.9%-Gd) (REO). MFCD00149731. Molecular Weight: 263.61. Molecular Formula: GdCl3∙XH2O. Color/form: white pwdr. Strem# 93-6421. http://www.strem.com/catalog/v/93-6421/
Catalog No. 50-901-16063 Supplier Strem, An Ascensus Company Supplier No. 936421100G
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
CAS# 19423-81-5. 100g. Gadolinium(III) chloride hydrate (99.9%-Gd) (REO). MFCD00149731. Molecular Weight: 263.61. Molecular Formula: GdCl3∙XH2O. Color/form: white pwdr. Strem# 93-6421. http://www.strem.com/catalog/v/93-6421/