Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gadolinium(III) Chloride Solution NMR Reference Standard, 0.1 mg/mL in D2O (99.9 atom % D), MilliporeSigma™ Supelco™

Catalog No. 111002893 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 x 0.235 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2893 10 x 0.235 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2893 Supplier MilliporeSigma Supelco Supplier No. 61273110X8IN
Add to Cart
Add to Cart

NMR Tube Size: 3 mm 8 in.

TRUSTED_SUSTAINABILITY

Specifications

Packaging NMR Tube Size: 3 mm 8 in.
Concentration 0.1 mg/mL in D2O (99.9 atom% D)
Linear Formula GdCl3
MDL Number MFCD00011024
Quantity 10 x 0.235 mL
Formula Weight 263.61
Grade NMR Reference Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.