Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gadolinium(III) chloride, ultra dry, 99.99% (REO)

Catalog No. AA3573614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3573614 25 g
AA3573606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3573614 Supplier Thermo Scientific Chemicals Supplier No. 03573614
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 10138-52-0 | Cl3Gd | 263.6 g/mol

Gadolinium(III) Chloride is used as chelating agents, which is essential for biomedical applications. It is also employed in the synthesis and nuclear magnetic resonance (NMR) studies of paramagnetic GdPCTA-[12] and GdPCTA-[13] complexes. It plays an important role as contrast agent for magnetic resonance imaging and an macrophage inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10138-52-0
Molecular Formula Cl3Gd
Molecular Weight (g/mol) 263.6
MDL Number MFCD00011024
InChI Key MEANOSLIBWSCIT-UHFFFAOYSA-K
Synonym gadolinium chloride, gadolinium trichloride, gadolinium chloride gdcl3, gdcl3, gadolinium iii chloride, gadolinium 3+ chloride, wln: gd g3, gadolinium iii chloride, ultra dry, gadolinium iii chloride, anhydrous, ?gdcl3
PubChem CID 61486
ChEBI CHEBI:37288
IUPAC Name trichlorogadolinium
SMILES Cl[Gd](Cl)Cl

Specifications

Melting Point 609°C
Color White
Physical Form Powder
Quantity 25 g
Sensitivity Hygroscopic
Appearance White powder
Assay Percent Range 99.99% (REO)
Solubility Information Soluble in water.
Formula Weight 263.61
Chemical Name or Material Gadolinium(III) chloride
Hazard Category H315-H319-H335-H500
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-386-6
RTECSNumber LW4025000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.