Learn More
Strem, An Ascensus Company CAS# 13450-87-8. 100g. Gadolinium(III) sulfate octahydrate (99.9%-Gd) (REO). MFCD00149734
SDP
Supplier:  Strem, An Ascensus Company 936407100G
Encompass
CAS# 13450-87-8. 100g. Gadolinium(III) sulfate octahydrate (99.9%-Gd) (REO). MFCD00149734. Molecular Weight: 602.68 (746.81). Molecular Formula: Gd2(SO4)3∙8H2O. Color/form: white xtl. Strem# 93-6407. http://www.strem.com/catalog/v/93-6407/
Catalog No. 50-901-16057