Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gadolinium(III) sulfate octahydrate, REacton™, 99.9% (REO)

Catalog No. AA4286918
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4286918 50 g
AA4286909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4286918 Supplier Thermo Scientific Chemicals Supplier No. 04286918
Add to Cart

missing translation for 'validMainframeMsg'

Cryogenic research | CAS: 13450-87-8 | Gd2H16O20S3 | 746.788 g/mol

Gadolinium(III) sulfate is used as analytical reagents, in inorganic synthesis and electronic ceramics. It is also useful in cryogenic research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13450-87-8
Molecular Formula Gd2H16O20S3
Molecular Weight (g/mol) 746.788
MDL Number MFCD00149734
InChI Key JBFCVGZGWFIPNK-UHFFFAOYSA-H
Synonym gadolinium iii sulfate octahydrate, digadolinium 3+ octahydrate trisulfate, gadolinium iii sulfate, gadolinium sulfate octahydrate, gadolinium sulfuric acid octahydrate, gadolinium sulfate, gadolinium sulfate-water 2/3/8, gadolinium iii sulfate octahydrate, reacton, gadolinium iii sulfate octahydrate trace metals basis
PubChem CID 16211473
IUPAC Name gadolinium(3+);trisulfate;octahydrate
SMILES O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Gd+3].[Gd+3]

Specifications

Physical Form Crystalline Aggregates
Quantity 50 g
Odor Odorless
Density 4.139 g/mL
Merck Index 14,4326
Solubility Information Soluble in cold water
Formula Weight 746.81 (602.68 Anhydrous)
Percent Purity 99.9%
Grade Reacton
Assay (REO)
Chemical Name or Material Gadolinium (III) sulfate octahydrate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 237-115-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.