Learn More
Medchemexpress LLC HY-A0167 50mg Medchemexpress, Gadopentetate (dimeglumine) CAS:86050-77-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYA016750MG
Encompass_Preferred
Medchemexpress, HY-A0167 50mg Gadopentetate (dimeglumine) CAS:86050-77-3 Gadopentetate dimeglumine (SH-L-451A) is used in combination with magnetic resonance imaging (MRI) to allow blood vessels, organs, and other non-bony tissues to be seen more clearly on the MRI. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2331