Learn More
Selleck Chemical LLC Gadopentetate Dimeglumine S4852-25mg
SDP
Supplier:  Selleck Chemical LLC S485225MG
Encompass_Preferred
Gadopentetate Dimeglumine (Gadopentetic acid dimeglumine Gd-DTPA Meglumine gadopentetate) a complex of gadolinium with a chelating agent diethylenetriamine penta-acetic acid is an extracellular intravenous contrast agent used in magnetic resonance imaging
Catalog No. 50-311-1373