Learn More
Medchemexpress LLC HY-107353 100mg Medchemexpress, Gadopentetic acid CAS:80529-93-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY107353100MG
Encompass_Preferred
Medchemexpress, HY-107353 100mg Gadopentetic acid CAS:80529-93-7 Gadopentetic acid (Gd-DTPA) is an paramagnetic contrast agent commonly implemented by a bolus intravenous injection (i.v.) in Dynamic contrast-enhanced MRI (DCE-MRI) studies. The initial concentration of Gadopentetic acid (Gd-DTPA) in the plasma ([Gd-DTPA 0 ]) is an important parameter for DCE-MRI. [Gd-DTPA 0 ] is related to the administered bolus dose and varies with subjects. A bolus of 0.025 mmol/kg Gd-DTPA offers shorter acquisition time and less exposure of subjects, with a half-life of 37.3 mins, a mean residence time of 53.8 mins, and an AUC of 3.37 ± 0.47 mmol/min/L [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2281