Learn More
MSC Tape Logic Gaffers Tape, 3 in x 60 yd, 16/Pk.
Catalog No. 501919276
Encompass
Industrial grade gaffers tape designed for securing wires to various surfaces.
  • Will not leave a residue
  • Suitable for indoor and outdoor use
  • Non-reflective
  • Provides 36 lb/in tensile strength
Catalog No. 50-191-9276 Supplier MSC Supplier No. 39588892
Industrial grade gaffers tape designed for securing wires to various surfaces.
  • Will not leave a residue
  • Suitable for indoor and outdoor use
  • Non-reflective
  • Provides 36 lb/in tensile strength