Learn More
MSC PEC Tools Screw Pitch Gage, Steel, 30 Blades, 1 ea
Catalog No. 5000516792
Encompass
This screw pitch gage is designed for the rapid and accurate identification of thread pitches. Featuring 30 blades, this tool is suitable for determining the pitch of both internal and external threads due to its narrow design. The set includes a sturdy steel protective case with locking devices at each end to secure blades during storage or to hold an individual blade in position during use.
  • Features 30 blades for a comprehensive pitch range of 6 to 60 threads per in.
  • Constructed from high-quality steel for durability.
  • Narrow blade profile facilitates use on internal and external threads.
  • Includes a protective steel case with dual locking devices.
  • Individual blades are clearly marked with thread sizes for easy identification.
Catalog No. 50-005-16792 Supplier MSC Supplier No. 06377220
This screw pitch gage is designed for the rapid and accurate identification of thread pitches. Featuring 30 blades, this tool is suitable for determining the pitch of both internal and external threads due to its narrow design. The set includes a sturdy steel protective case with locking devices at each end to secure blades during storage or to hold an individual blade in position during use.
  • Features 30 blades for a comprehensive pitch range of 6 to 60 threads per in.
  • Constructed from high-quality steel for durability.
  • Narrow blade profile facilitates use on internal and external threads.
  • Includes a protective steel case with dual locking devices.
  • Individual blades are clearly marked with thread sizes for easy identification.