Learn More
MSC Value Collection, Plug Gage Handle Bushing, 0.11 to 0.117"
Catalog No. 5000510108
Encompass
This plug gage handle bushing is designed for use in precision measurement applications, providing a secure fit for plug gages within the specified size range. It is suitable for inspecting and testing environments requiring accurate go/no go gaging.
  • Accommodates gages from 0.110 in. to 0.117 in.
  • Constructed for reliability in metalworking and inspection workflows
  • Engineered for consistent performance during gage mounting
Catalog No. 50-005-10108 Supplier MSC Supplier No. 88800156
This plug gage handle bushing is designed for use in precision measurement applications, providing a secure fit for plug gages within the specified size range. It is suitable for inspecting and testing environments requiring accurate go/no go gaging.
  • Accommodates gages from 0.110 in. to 0.117 in.
  • Constructed for reliability in metalworking and inspection workflows
  • Engineered for consistent performance during gage mounting