Learn More
MSC Plug Gage Handle Bushing
Catalog No. 5000516445
Encompass
This plug gage handle bushing is designed for use in precision measurement applications, specifically accommodating plug gages within a specified diameter range. It provides a reliable interface for secure tool handling during inspection processes.
  • Designed for plug gage handles
  • Measurement range of 0.244 in. to 0.252 in.
  • Constructed for industrial use
Catalog No. 50-005-16445 Supplier MSC Supplier No. 88800339
This plug gage handle bushing is designed for use in precision measurement applications, specifically accommodating plug gages within a specified diameter range. It provides a reliable interface for secure tool handling during inspection processes.
  • Designed for plug gage handles
  • Measurement range of 0.244 in. to 0.252 in.
  • Constructed for industrial use