Learn More
MSC Value Collection, Plug Gage Handle Bushing, 0.781 to 0.812"
Catalog No. 5000510515
Encompass
This plug gage handle bushing is designed for use in Go/No Go inspection processes, facilitating secure mounting of plug gages within a handle. It accommodates gage diameters ranging from 0.781 in. to 0.812 in.
  • Fits gages with diameters from 0.781 in. to 0.812 in.
  • Constructed for reliable use in inspection applications
  • Facilitates secure assembly of plug gages into handle units
Catalog No. 50-005-10515 Supplier MSC Supplier No. 88800735
This plug gage handle bushing is designed for use in Go/No Go inspection processes, facilitating secure mounting of plug gages within a handle. It accommodates gage diameters ranging from 0.781 in. to 0.812 in.
  • Fits gages with diameters from 0.781 in. to 0.812 in.
  • Constructed for reliable use in inspection applications
  • Facilitates secure assembly of plug gages into handle units