Learn More
Grainger Gage Pin Set ZZ .006 -.06 55 pcs.
Catalog No. 502751176
Encompass_Preferred
The Grainger Approved Gage Pin Set is a set of precision ground steel pins used for calibrating measuring instruments and setting up precision assemblies. The set contains 9 pins ranging from 0.4990 to 1.4995 inches in 0.001 inch increments. The pins are made from hardened and ground alloy steel for durability and accuracy. They come in a fitted plastic case for storage and portability. The gage pins can be used to calibrate inside, outside, and depth micrometers. They are an essential inspection and measurement tool for any machine shop or inspection area.
Catalog No. 50-275-1176 Supplier Grainger Supplier No. 141F58
The Grainger Approved Gage Pin Set is a set of precision ground steel pins used for calibrating measuring instruments and setting up precision assemblies. The set contains 9 pins ranging from 0.4990 to 1.4995 inches in 0.001 inch increments. The pins are made from hardened and ground alloy steel for durability and accuracy. They come in a fitted plastic case for storage and portability. The gage pins can be used to calibrate inside, outside, and depth micrometers. They are an essential inspection and measurement tool for any machine shop or inspection area.