Learn More
Grainger Gage Pin Set ZZ .011 -.25 240 pcs.
Catalog No. 502753020
Encompass_Preferred
The Grainger Approved Gage Pin Set is a set of precision ground steel pins used for calibrating measuring instruments. The set contains 9 pins ranging from 0.100" to 1.000" in 0.100" increments, with a tolerance of +/- 0.0002". The pins have a knurled grip area and are made from hardened and tempered alloy steel. They come in a fitted plastic case for storage and portability. The pins allow for accurate calibration and inspection of gages, micrometers, dial indicators and other precision measuring tools. This gage pin set provides an economical solution for workshop calibration needs.
Catalog No. 50-275-3020 Supplier Grainger Supplier No. 141F60
The Grainger Approved Gage Pin Set is a set of precision ground steel pins used for calibrating measuring instruments. The set contains 9 pins ranging from 0.100" to 1.000" in 0.100" increments, with a tolerance of +/- 0.0002". The pins have a knurled grip area and are made from hardened and tempered alloy steel. They come in a fitted plastic case for storage and portability. The pins allow for accurate calibration and inspection of gages, micrometers, dial indicators and other precision measuring tools. This gage pin set provides an economical solution for workshop calibration needs.