Learn More
Grainger Gage Pin Set ZZ Minus 1.01mm-13.99mm
Catalog No. 502752381
Encompass_Preferred
The Grainger Approved Gage Pin Set - ZZ Minus 1.01mm - 13.99mm is a precision measuring tool used to calibrate measuring instruments and check tolerances. This set contains 13 gage pins with diameters ranging from 1.01mm to 13.99mm, with each pin diameter decreasing by 1mm. The gage pins are made from hardened and ground tool steel with a tolerance of +/- .0001mm. They feature a knurled grip area and come in a fitted plastic case for storage and organization. This gage pin set provides a wide range of diameters for versatile use in calibration and precision measurement.
Catalog No. 50-275-2381 Supplier Grainger Supplier No. 141H22
The Grainger Approved Gage Pin Set - ZZ Minus 1.01mm - 13.99mm is a precision measuring tool used to calibrate measuring instruments and check tolerances. This set contains 13 gage pins with diameters ranging from 1.01mm to 13.99mm, with each pin diameter decreasing by 1mm. The gage pins are made from hardened and ground tool steel with a tolerance of +/- .0001mm. They feature a knurled grip area and come in a fitted plastic case for storage and organization. This gage pin set provides a wide range of diameters for versatile use in calibration and precision measurement.